Ethyl formate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Formic acid ethyl ester

A11113-0F A11113-AE A11113-AP
CAAAA11113-0FEA 202.24 CAD
CAAAA11113-0F CAAAA11113-AE CAAAA11113-AP
Ethyl formate 97%
Ethyl formate
Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 148
Melting Pt: -2
Density: 0.921
Flash Pt: –19 °C (–2 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00003294
CAS Number: 109-94-4
EINECS: 203-721-0
UN: 1190
ADR: 3 II
Merck Index: 14,3807

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR