Veratraldehyde 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,4-Dimethoxybenzaldehyde

A11110-0E A11110-22 A11110-36
CAAAA11110-0EEA 592.5 CAD
CAAAA11110-0E CAAAA11110-22 CAAAA11110-36
Veratraldehyde 99%
Veratraldehyde
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 281 °C
Melting Pt: 42…46 °C
Flash Pt: 112 °C(233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003363
CAS Number: 120-14-9
EINECS: 204-373-2
Merck Index: 14,9951

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR