1,3-Cyclohexanedione 97+% may cont. up to 1% NaCl

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclohexane-1,3-dione

A11109-14 A11109-22 A11109-0B A11109-18
CAAAAA11109-14EA 42.66 CAD
CAAAAA11109-14 CAAAAA11109-22 CAAAA11109-0B CAAAA11109-18
1,3-Cyclohexanedione 97+% may cont. up to 1% NaCl
1,3-Cyclohexanedione

MDL: MFCD00001585 Beilstein Registry No.: 385888 Fieser: 1,178


Purity: 97+% CAS No.: 504-02-9 Molecular Formula: C6H8O2 Formula Weight: 112.13 Storage Temperature: 2°C to 8°C Melting Point: 101-105° Beilstein Reference: 385888 Merck Reference: 14,3178 MDL No.: MFCD00001585

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 235 °C (1013 hPa)
Melting Pt: 101…105 °C
Density: 1.100
Storage Temperature: Refrigerator
MDL Number: MFCD00001585
CAS Number: 504-02-9
EINECS: 207-980-0
Merck Index: 14,3178

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR