4-Vinylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11107-03
A11107-06
A11107-14
CAAAA11107-03EA
184.86
CAD
CAAAA11107-03
CAAAA11107-06
CAAAA11107-14
4-Vinylbenzoic acid 98%
4-Vinylbenzoic acid
MDL: MFCD00002569
Beilstein Registry No.: 2041130
Purity: 98%
CAS No.: 1075-49-6
Molecular Formula: C9H8O2
Formula Weight: 148.16
Storage Temperature: 2°C to 8°C
Melting Point: ca 142°
Beilstein Reference: 2041130
MDL No.: MFCD00002569
Formula:
C₉H₈O₂ Boiling Pt: 282 °C (1013 hPa) Melting Pt: ∼ 142 °C Density: 1.30 Storage Temperature: Refrigerator |
MDL Number:
MFCD00002569 CAS Number: 1075-49-6 EINECS: 214-053-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...