(4-Nitrophenyl)acetic acid ethyl ester 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 4-nitrohenylacetate, Ethyl (4-nitrophenyl)acetate, Ethyl 4-nitrophenylacetate

A11101-06 A11101-14 A11101-22
CAAAA11101-06EA 92.59 CAD
CAAAA11101-06 CAAAA11101-14 CAAAA11101-22
(4-Nitrophenyl)acetic acid ethyl ester 98%
(4-Nitrophenyl)acetic acid ethyl ester

MDL: MFCD00017046 Beilstein Registry No.: 2053241


Purity: 98% CAS No.: 5445-26-1 Molecular Formula: C10H11NO4 Formula Weight: 209.20 Melting Point: 62-66° Beilstein Reference: 2053241

Formula: C₁₀H₁₁NO₄
MW: 209.2 g/mol
Melting Pt: 62…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00017046
CAS Number: 5445-26-1
EINECS: 226-646-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR