1,4-Cyclohexanedione 98%
Supplier: Thermo Scientific Chemicals
A11100-36
A11100-14
A11100-22
CAAAAA11100-36EA
1429.9
CAD
CAAAAA11100-36
CAAAA11100-14
CAAAA11100-22
1,4-Cyclohexanedione 98%
1,4-Cyclohexanedione
MDL: MFCD00001606
Beilstein Registry No.: 774152
Fieser: 1,178
Purity: 98%
CAS No.: 637-88-7
Molecular Formula: C6H8O2
Formula Weight: 112.13
Storage Temperature: 2°C to 8°C
Flash Point: 132°(269°F)
Melting Point: 77-78°
Boiling Point: 132°/20mm
Beilstein Reference: 774152
MDL No.: MFCD00001606
Formula:
C₆H₈O₂ Boiling Pt: 132 °C/20 mm Melting Pt: 77…78 °C Density: 1.28 g/cm³ (20 °C) Flash Pt: 132 °C(269 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001606 CAS Number: 637-88-7 EINECS: 211-306-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...