Tris(2-methoxyphenyl)phosphine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tris(o-methoxyphenyl)phosphine

A11096-03 A11096-06
CAAAAA11096-14EA 1201.31 CAD
CAAAAA11096-14 CAAAA11096-03 CAAAA11096-06
Tris(2-methoxyphenyl)phosphine 98%
Tris(2-methoxyphenyl)phosphine

MDL: MFCD00014892 Beilstein Registry No.: 2776186


Purity: 98% CAS No.: 4731-65-1 Molecular Formula: C21H21O3P Formula Weight: 352.37 Melting Point: 196-204° Beilstein Reference: 2776186

Formula: C₂₁H₂₁O₃P
MW: 352.37 g/mol
Melting Pt: 196…204 °C
Storage Temperature: Ambient
MDL Number: MFCD00014892
CAS Number: 4731-65-1
EINECS: 225-235-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR