2-Ethyl-3-methylpyrazine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11092-06
A11092-14
A11092-22
CAAAA11092-06EA
55.62
CAD
CAAAA11092-06
CAAAA11092-14
CAAAA11092-22
2-Ethyl-3-methylpyrazine 98+%
2-Ethyl-3-methylpyrazine
MDL: MFCD00006150
Beilstein Registry No.: 956775
Purity: 98+%
CAS No.: 15707-23-0
Molecular Formula: C7H10N2
Formula Weight: 122.17
Flash Point: 57°(134°F)
Refractive Index: 1.5030
Odor: Characteristic
Boiling Point: 169-171°
Beilstein Reference: 956775
UN No.: 1993
MDL No.: MFCD00006150
Formula:
C₇H₁₀N₂ Boiling Pt: 169…171 °C Density: 0.987 Flash Pt: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006150 CAS Number: 15707-23-0 EINECS: 239-799-8 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...