D-(+)-Glucose monohydrate

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate

A11090-0E A11090-36
CAAAA11090-0EEA 197.5 CAD
CAAAA11090-0E CAAAA11090-36
D-(+)-Glucose monohydrate
D(+)-Glucose monohydrate

MDL: MFCD00149450


CAS No.: 14431-43-7 Molecular Formula: C6H12O6.H2O Formula Weight: 198.18 (180.16anhy) Physical Form: Cryst powder Appearance: White Melting Point: ca 83° MDL No.: MFCD00149450

Formula: C₆H₁₂O₆·H₂O
MW: 198.17 g/mol
Melting Pt: ∼ 83 °C
Storage Temperature: Ambient
MDL Number: MFCD00149450
CAS Number: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR