Phenazone 98%

Supplier: Thermo Scientific Chemicals
Warning

A11089-22 A11089-36
CAAAA11089-22EA 53.09 CAD
CAAAA11089-22 CAAAA11089-36
Phenazone 98%
Phenazone

MDL: MFCD00003146 Beilstein Registry No.: 157775


Purity: 98% CAS No.: 60-80-0 Molecular Formula: C11H12N2O Formula Weight: 188.23 Melting Point: 111-114° Beilstein Reference: 157775 Merck Reference: 14,716 MDL No.: MFCD00003146

Formula: C₁₁H₁₂N₂O
Boiling Pt: 388 °C (1013 hPa)
Melting Pt: 111…114 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003146
CAS Number: 60-80-0
EINECS: 200-486-6
Merck Index: 14,716

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR