5-Chlorouracil 98%

Supplier: Thermo Scientific Chemicals

A11084-06 A11084-14
CAAAA11084-06EA 112.02 CAD
CAAAA11084-06 CAAAA11084-14 CAAAA11084-22
5-Chlorouracil 98%
5-Chlorouracil

MDL: MFCD00006019 Beilstein Registry No.: 127173


Purity: 98% CAS No.: 1820-81-1 Molecular Formula: C4H3ClN2O2 Formula Weight: 146.53 Melting Point: 300° Beilstein Reference: 127173

Formula: C₄H₃ClN₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006019
CAS Number: 1820-81-1
EINECS: 217-339-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR