Sodium butyrate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Butyric acid sodium salt, Sodium Butyrate

A11079-06 A11079-22 A11079-36
CAAAAA11079-06EA 47.08 CAD
CAAAAA11079-06 CAAAA11079-22 CAAAA11079-36
Sodium butyrate 98+%
Sodium butyrate

Decreases calcium release from intracellular stores. Inhibits histone deacetylase


Purity: 98+% CAS No.: 156-54-7 Molecular Formula: C4H7NaO2 Formula Weight: 110.09 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Melting Point: 300° Beilstein Reference: 3629439 MDL No.: MFCD00002816


Decreases calcium release from intracellular stores. Inhibits histone deacetylase

Formula: C₄H₇NaO₂
MW: 110.09 g/mol
Melting Pt: >300 °C
Density: 1.324 g/cm³ (30 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002816
CAS Number: 156-54-7
EINECS: 205-857-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR