3-Acetylthiophene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11078-09
A11078-18
A11078-30
CAAAA11078-09EA
167.48
CAD
CAAAA11078-09
CAAAA11078-18
CAAAA11078-30
3-Acetylthiophene 98%
3-Acetylthiophene
MDL: MFCD00005468
Beilstein Registry No.: 107241
Purity: 98%
CAS No.: 1468-83-3
Molecular Formula: C6H6OS
Formula Weight: 126.18
Storage Temperature: 2°C to 8°C
Melting Point: 58-61°
Boiling Point: 208-210°
Beilstein Reference: 107241
UN No.: 2811
MDL No.: MFCD00005468
Formula:
C₆H₆OS Boiling Pt: 208…210 °C Melting Pt: 58…61 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005468 CAS Number: 1468-83-3 EINECS: 215-996-4 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...