Xanthine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11077-14
A11077-22
CAAAA11077-14EA
132.72
CAD
CAAAA11077-14
CAAAA11077-22
Xanthine 99%
Xanthine
MDL: MFCD00078453
Beilstein Registry No.: 8733
Purity: 99%
CAS No.: 69-89-6
Molecular Formula: C5H4N4O2
Formula Weight: 152.11
Odor: Odorless
Melting Point:
Formula:
C₅H₄N₄O₂ Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078453 CAS Number: 69-89-6 EINECS: 200-718-6 Merck Index: 14,10059 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...