Formamide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Carbamaldehyde
A11076-0C
A11076-0E
A11076-36
CAAAA11076-0CEA
567.22
CAD
CAAAA11076-0C
CAAAA11076-0E
CAAAA11076-36
Formamide 99%
Formamide
MDL: MFCD00007941
Purity: 99%
CAS No.: 75-12-7
Molecular Formula: CH3NO
Formula Weight: 45.04
Flash Point: 150°(302°F)
Refractive Index: 1.4475
Odor: Ammonia-like
Melting Point: 2°
Boiling Point: 210°
Beilstein Reference: 505995
Merck Reference: 14,4237
MDL No.: MFCD00007941
Formula:
HCONH₂ MW: 45.04 g/mol Boiling Pt: 210 °C Melting Pt: 2 °C Density: 1.134 Flash Pt: 150 °C(302 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007941 CAS Number: 75-12-7 EINECS: 200-842-0 Merck Index: 14,4237 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...