DL-Lysine monohydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-2,6-Diaminocaproic acid monohydrochloride, H-DL-Lys-OH · HCl, H-DL-Lys-OH.HCl, (±)-2,6-Diaminohexanoic acid monohydrochloride, (±)-Lysine monohydrochloride

A11066-14 A11066-22
CAAAA11066-14EA 58.62 CAD
CAAAA11066-14 CAAAA11066-22
DL-Lysine monohydrochloride 99%
DL-Lysine monohydrochloride

MDL: MFCD00064563 Beilstein Registry No.: 4711993


Purity: 99% CAS No.: 70-53-1 Molecular Formula: C6H14N2O2.HCl Formula Weight: 182.65 Melting Point: 267° dec. Beilstein Reference: 4711993

Formula: C₆H₁₅ClN₂O₂
MW: 182.65 g/mol
Melting Pt: 267 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064563
CAS Number: 70-53-1
EINECS: 200-739-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR