6-Bromohexanoyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A11063-06 A11063-14 A11063-22
CAAAAA11063-06EA 51.82 CAD
CAAAAA11063-06 CAAAA11063-14 CAAAA11063-22 CAAAA11063-36
6-Bromohexanoyl chloride 97%
6-Bromohexanoyl chloride

MDL: MFCD00000761 Beilstein Registry No.: 1749739


Purity: 97% CAS No.: 22809-37-6 Molecular Formula: C6H10BrClO Formula Weight: 213.50 Flash Point: 110°(230°F) Refractive Index: 1.4875 Odor: Acrid Boiling Point: 119-120°/9mm Beilstein Reference: 1749739 UN No.: 3265 MDL No.: MFCD00000761

Formula: BrH₂C(CH₂)₄COOCl
Boiling Pt: 119…120 °C/9 mm
Density: 1.397
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000761
CAS Number: 22809-37-6
EINECS: 245-236-7
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR