1-Fluoro-4-nitrobenzene 99%

Supplier: Thermo Scientific Chemicals
Warning

A11057-0E A11057-22 A11057-36
CAAAA11057-0EEA 1020.68 CAD
CAAAA11057-0E CAAAA11057-22 CAAAA11057-36
1-Fluoro-4-nitrobenzene 99%
1-Fluoro-4-nitrobenzene

MDL: MFCD00007282 Beilstein Registry No.: 606923


Purity: 99% CAS No.: 350-46-9 Molecular Formula: C6H4FNO2 Formula Weight: 141.10 Flash Point: 83°(181°F) Refractive Index: 1.5315 Melting Point: 22-24° Boiling Point: 204-206° Beilstein Reference: 606923 UN No.: 2811 MDL No.: MFCD00007282

Formula: C₆H₄FNO₂
Boiling Pt: 204…206 °C
Melting Pt: 22…24 °C
Density: 1.330
Flash Pt: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007282
CAS Number: 350-46-9
EINECS: 206-502-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR