1-Indanone 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11056-06
A11056-14
A11056-22
A11056-36
CAAAAA11056-06EA
49.45
CAD
CAAAAA11056-06
CAAAA11056-14
CAAAA11056-22
CAAAA11056-36
1-Indanone 99+%
1-Indanone
MDL: MFCD00003785
Beilstein Registry No.: 507957
Purity: 99+%
CAS No.: 83-33-0
Molecular Formula: C9H8O
Formula Weight: 132.16
Flash Point: 113°(235°F)
Melting Point: 38-42°
Boiling Point: 243-245°
Beilstein Reference: 507957
MDL No.: MFCD00003785
Formula:
C₉H₈O Boiling Pt: 243…245 °C Melting Pt: 38…42 °C Density: 1.103 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003785 CAS Number: 83-33-0 EINECS: 201-470-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...