Cyanoacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11045-0B
A11045-0I
A11045-30
CAAAA11045-0BEA
175.38
CAD
CAAAA11045-0B
CAAAA11045-0I
CAAAA11045-30
Cyanoacetic acid 97%
Cyanoacetic acid
MDL: MFCD00002677
Beilstein Registry No.: 506325
Fieser: 11,145
Purity: 99%
CAS No.: 372-09-8
Molecular Formula: NCCH2CO2H
Formula Weight: 85.06
Flash Point: 107°(224°F)
Odor: Odorless
pH: 1.5
Melting Point: 64-69°
Boiling Point: 108°/15mm
Beilstein Reference: 506325
Merck Reference: 14,2689
UN No.: 3261
MDL No.: MFCD00002677
Formula:
C₃H₃NO₂ MW: 85.06 g/mol Boiling Pt: 108 °C/15 mm Melting Pt: 64…69 °C Density: Min. 1.1 g/cm³ (20 °C) Flash Pt: 107 °C (224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002677 CAS Number: 372-09-8 EINECS: 206-743-9 UN: 3261 ADR: 8,III Merck Index: 14,2689 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...