2-Bromo-4-fluoroanisole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11040-09
A11040-18
CAAAA11040-09EA
49.45
CAD
CAAAA11040-09
CAAAA11040-18
2-Bromo-4-fluoroanisole 98%
2-Bromo-4-fluoroanisole
MDL: MFCD00012014
Beilstein Registry No.: 2358880
Purity: 98%
CAS No.: 452-08-4
Molecular Formula: C7H6BrFO
Formula Weight: 205.03
Flash Point: 98°(208°F)
Refractive Index: 1.5450
Boiling Point: 88-89°/1mm
Beilstein Reference: 2358880
MDL No.: MFCD00012014
Formula:
C₇H₆BrFO Boiling Pt: 88…89 °C/1 mm Melting Pt: –27 °C Density: 1.581 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00012014 CAS Number: 452-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...