Diethylenetriamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2,2'-Iminodi(ethylamine), DIEN, Diethylenetriamine
A11033-AE
A11033-0F
A11033-AP
CAAAAA11033-AEEA
60.99
CAD
CAAAAA11033-AE
CAAAA11033-0F
CAAAA11033-AP
Diethylenetriamine 99%
Diethylenetriamine
MDL: MFCD00008171
Beilstein Registry No.: 605314
Purity: 99%
CAS No.: 111-40-0
Molecular Formula: C4H13N3
Formula Weight: 103.17
Flash Point: 94°(201°F)
Refractive Index: 1.4830
Odor: Ammonia-like
Melting Point: -40°
Boiling Point: 200-204°
Beilstein Reference: 605314
UN No.: 2079
MDL No.: MFCD00008171
Formula:
C₄H₁₃N₃ Boiling Pt: 200…204 °C Melting Pt: –40 °C Density: 0.952 Flash Pt: 94 °C (201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008171 CAS Number: 111-40-0 EINECS: 203-865-4 UN: 2079 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...