4'-Ethoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11031-06
A11031-14
A11031-22
CAAAA11031-06EA
48.98
CAD
CAAAA11031-06
CAAAA11031-14
CAAAA11031-22
4'-Ethoxyacetophenone 98%
4'-Ethoxyacetophenone
MDL: MFCD00009095
Beilstein Registry No.: 636783
Purity: 98%
CAS No.: 1676-63-7
Molecular Formula: C10H12O2
Formula Weight: 164.20
Flash Point: 134°(273°F)
Melting Point: 35-38°
Boiling Point: 145-146°/13mm
Beilstein Reference: 636783
MDL No.: MFCD00009095
Formula:
C₁₀H₁₂O₂ Boiling Pt: 145…146 °C/13 mm Melting Pt: 35…38 °C Flash Pt: 134 °C(273 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009095 CAS Number: 1676-63-7 EINECS: 216-825-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...