4-Chlorothioanisole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulfide, 4-Chlorophenyl methyl sulphide

A11025-06 A11025-14
CAAAA11025-06EA 69.2 CAD
CAAAA11025-06 CAAAA11025-14
4-Chlorothioanisole 98%
4-Chlorothioanisole

MDL: MFCD00013643 Beilstein Registry No.: 2041509

Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 223 °C
Melting Pt: 27…28 °C
Density: 1.220
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013643
CAS Number: 123-09-1
EINECS: 204-600-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR