2-Bromo-4,6-difluoroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11022-14
A11022-22
A11022-36
CAAAA11022-14EA
104.6
CAD
CAAAA11022-14
CAAAA11022-22
CAAAA11022-36
2-Bromo-4,6-difluoroaniline 98%
2-Bromo-4,6-difluoroaniline
MDL: MFCD00009639
Beilstein Registry No.: 2718139
Purity: 98%
CAS No.: 444-14-4
Molecular Formula: C6H4BrF2N
Formula Weight: 208.01
Odor: Weak, characteristic
Melting Point: 38-42°
Beilstein Reference: 2718139
UN No.: 3077
MDL No.: MFCD00009639
Formula:
C₆H₄BrF₂N Boiling Pt: 227 °C (1013 hPa) Melting Pt: 38…42 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009639 CAS Number: 444-14-4 EINECS: 429-430-0 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...