Isobutylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Amino-2-methylpropane, 2-Methylpropylamine
A11017-AK
A11017-AU
CAAAA11017-AKEA
46.29
CAD
CAAAA11017-AK
CAAAA11017-AU
Isobutylamine 99%
Isobutylamine
MDL: MFCD00008146
Beilstein Registry No.: 385626
Purity: 99%
CAS No.: 78-81-9
Molecular Formula: (CH3)2CHCH2NH2
Formula Weight: 73.14
Flash Point: -9°(15°F)
Refractive Index: 1.3970
Odor: Amine-like
Melting Point: -85°
Boiling Point: 67-69°
Beilstein Reference: 385626
Merck Reference: 14,5132
UN No.: 1214
MDL No.: MFCD00008146
Formula:
C₄H₁₁N MW: 73.14 g/mol Boiling Pt: 67…69 °C Melting Pt: –85 °C Density: 0.736 Flash Pt: –9 °C (15 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008146 CAS Number: 78-81-9 EINECS: 201-145-4 UN: 1214 ADR: 3,II Merck Index: 14,5132 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...