4-Aminobutyric acid 97%
Supplier: Thermo Scientific Chemicals
A11016-0E
A11016-22
A11016-36
CAAAA11016-0EEA
1333.52
CAD
CAAAA11016-0E
CAAAA11016-22
CAAAA11016-36
4-Aminobutyric acid 97%
4-Aminobutyric acid
MDL: MFCD00008226
Beilstein Registry No.: 906818
Purity: 97%
CAS No.: 56-12-2
Molecular Formula: C4H9NO2
Formula Weight: 103.12
Odor: Odorless
Melting Point: ca 195° dec.
Beilstein Reference: 906818
Merck Reference: 14,430
MDL No.: MFCD00008226
Formula:
C₄H₉NO₂ Boiling Pt: 248 °C (1013 hPa) Melting Pt: ∼ 195 °C (dec.) Density: 1.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00008226 CAS Number: 56-12-2 EINECS: 200-258-6 Merck Index: 14,430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...