2-Fluorobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11015-22 A11015-36
CAAAA11015-22EA 125.61 CAD
CAAAA11015-22 CAAAA11015-36
2-Fluorobenzoic acid 98+%
2-Fluorobenzoic acid

MDL: MFCD00002405 Beilstein Registry No.: 971265


Purity: 98+% CAS No.: 445-29-4 Molecular Formula: C7H5FO2 Formula Weight: 140.11 Melting Point: 122-125° Beilstein Reference: 971265

Formula: C₇H₅FO₂
Boiling Pt: 245 °C (1013 hPa)
Melting Pt: 122…125 °C
Density: 1,46
Storage Temperature: Ambient
MDL Number: MFCD00002405
CAS Number: 445-29-4
EINECS: 207-158-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR