α,α-Dimethoxytoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Benzaldehyde dimethyl acetal
A11009-0E
A11009-22
A11009-36
CAAAA11009-0EEA
1042.8
CAD
CAAAA11009-0E
CAAAA11009-22
CAAAA11009-36
α,α-Dimethoxytoluene 99%
α,α-Dimethoxytoluene
MDL: MFCD00008491
Beilstein Registry No.: 2044501
Purity: 99%
CAS No.: 1125-88-8
Molecular Formula: C9H12O2
Formula Weight: 152.20
Flash Point: 69°(156°F)
Refractive Index: 1.4935
Odor: Characteristic
Boiling Point: 196°
Beilstein Reference: 2044501
MDL No.: MFCD00008491
Formula:
C₉H₁₂O₂ MW: 152.19 g/mol Boiling Pt: 196 °C Density: 1.014 Flash Pt: 69 °C(156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008491 CAS Number: 1125-88-8 EINECS: 214-413-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...