Dimethyl malonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11007-0E A11007-22 A11007-36
CAAAA11007-0EEA 306.52 CAD
CAAAA11007-0E CAAAA11007-22 CAAAA11007-36
Dimethyl malonate 98+%
Dimethyl malonate

MDL: MFCD00008460 Beilstein Registry No.: 774261 Notes: Slightly soluble in water. Miscible with alcohol, ether, and oils Fieser: 8,193


Purity: 98+% CAS No.: 108-59-8 Molecular Formula: C5H8O4 Formula Weight: 132.12 Flash Point: 90°(194°F) Refractive Index: 1.4130 Melting Point: -62° Boiling Point: 180-181° Beilstein Reference: 774261 Merck Reference: 14,6096 MDL No.: MFCD00008460

Formula: C₅H₈O₄
Boiling Pt: 180…181 °C
Melting Pt: –62 °C
Density: 1.156
Flash Pt: 90 °C(194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008460
CAS Number: 108-59-8
EINECS: 203-597-8
Merck Index: 14,6096

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR