3,5-Dichlorobenzenesulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

A11003-03 A11003-06 A11003-14
CAAAA11003-03EA 43.29 CAD
CAAAA11003-03 CAAAA11003-06 CAAAA11003-14
3,5-Dichlorobenzenesulfonyl chloride 97%
3,5-Dichlorobenzenesulphonyl chloride

MDL: MFCD00051698 Beilstein Registry No.: 1528657


Purity: 97% CAS No.: 705-21-5 Molecular Formula: C6H3Cl3O2S Formula Weight: 245.51 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 158-160°/15mm Beilstein Reference: 1528657 UN No.: 3261 MDL No.: MFCD00051698

Formula: C₆H₃Cl₃O₂S
Boiling Pt: 158…160 °C/15 mm
Melting Pt: 30…33 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00051698
CAS Number: 705-21-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR