Indoline 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,3-Dihydroindol

A11000-0B A11000-18 A11000-30
CAAAA11000-0BEA 1153.4 CAD
CAAAA11000-0B CAAAA11000-18 CAAAA11000-30
Indoline 99%
Indoline

MDL: MFCD00005705 Beilstein Registry No.: 111915


Purity: 99% CAS No.: 496-15-1 Molecular Formula: C8H9N Formula Weight: 119.17 Flash Point: 92°(197°F) Refractive Index: 1.5930 Odor: Odorless Melting Point: -21° Boiling Point: 220-221° Beilstein Reference: 111915 MDL No.: MFCD00005705

Formula: C₈H₉N
MW: 119.17 g/mol
Boiling Pt: 220…221 °C
Melting Pt: –21 °C
Density: 1.062
Flash Pt: 92 °C(197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005705
CAS Number: 496-15-1
EINECS: 207-816-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR