3-Iodotoluene 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10999-22
A10999-18
A10999-30
CAAAAA10999-22EA
218.04
CAD
CAAAAA10999-22
CAAAA10999-18
CAAAA10999-30
3-Iodotoluene 99%
3-Iodotoluene
MDL: MFCD00001050
Beilstein Registry No.: 1903634
Purity: 99%
CAS No.: 625-95-6
Molecular Formula: C7H7I
Formula Weight: 218.04
Flash Point: 82°(179°F)
Refractive Index: 1.6040
Melting Point: -27°
Boiling Point: 80-82°/10mm
Beilstein Reference: 1903634
MDL No.: MFCD00001050
Formula:
C₇H₇I Boiling Pt: 80…82 °C/10 mm Melting Pt: –27 °C Density: 1.698 Flash Pt: 82 °C(179 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001050 CAS Number: 625-95-6 EINECS: 210-918-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...