Benzylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10997-36
A10997-0B
A10997-0I
A10997-30
CAAAAA10997-36EA
106.81
CAD
CAAAAA10997-36
CAAAA10997-0B
CAAAA10997-0I
CAAAA10997-30
Benzylamine 98+%
Benzylamine
MDL: MFCD00008106
Beilstein Registry No.: 741984
Fieser: 1,51 10,26 16,22
Purity: 98+%
CAS No.: 100-46-9
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 72°(162°F)
Refractive Index: 1.5435
Melting Point: -43°
Boiling Point: 184-185°
Beilstein Reference: 741984
Merck Reference: 14,1125
UN No.: 2735
MDL No.: MFCD00008106
Formula:
(C₆H₅)CH₂NH₂ Boiling Pt: 184…185 °C Melting Pt: –43 °C Density: 0.982 Flash Pt: 72 °C (162 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008106 CAS Number: 100-46-9 EINECS: 202-854-1 UN: 2735 ADR: 8,III Merck Index: 14,1125 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...