2-Phenylbenzimidazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10996-06
A10996-14
A10996-22
CAAAA10996-06EA
46.29
CAD
CAAAA10996-06
CAAAA10996-14
CAAAA10996-22
2-Phenylbenzimidazole 97%
2-Phenylbenzimidazole
MDL: MFCD00005592
Beilstein Registry No.: 7087
Purity: 97%
CAS No.: 716-79-0
Molecular Formula: C13H10N2
Formula Weight: 194.24
Melting Point: 296-297°
Beilstein Reference: 7087
Merck Reference: 14,7274
MDL No.: MFCD00005592
Formula:
C₁₃H₁₀N₂ Boiling Pt: 422 °C (1013 hPa) Melting Pt: 296…297 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005592 CAS Number: 716-79-0 EINECS: 211-939-2 Merck Index: 14,7274 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...