Methyl octanoate 99%
Supplier: Thermo Scientific Chemicals
Warning
|
A10991-18
A10991-30
CAAAA10991-0BEA
1786.24
CAD
CAAAA10991-0B
CAAAA10991-18
CAAAA10991-30
Methyl octanoate 99%
Methyl octanoate
MDL: MFCD00009551
Beilstein Registry No.: 1752270
Purity: 99%
CAS No.: 111-11-5
Molecular Formula: CH3(CH2)6CO2CH3
Formula Weight: 158.24
Flash Point: 72°(161°F)
Refractive Index: 1.4170
Melting Point: -40°
Boiling Point: 193-194°
Beilstein Reference: 1752270
MDL No.: MFCD00009551
Formula:
C₉H₁₈O₂ Boiling Pt: 193…194 °C Melting Pt: –40 °C Density: 0.875 Flash Pt: 72 °C(161 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009551 CAS Number: 111-11-5 EINECS: 203-835-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...