4-Bromo-1-butene 97%

Supplier: Thermo Scientific Chemicals
Danger

A10977-06 A10977-14
CAAAA10977-06EA 101.12 CAD
CAAAA10977-06 CAAAA10977-14
4-Bromo-1-butene 97%
4-Bromo-1-butene

MDL: MFCD00000258 Beilstein Registry No.: 1098377


Purity: 97% CAS No.: 5162-44-7 Molecular Formula: BrCH2CH2CH=CH2 Formula Weight: 135.00 Storage Temperature: 2°C to 8°C Flash Point: 1°(34°F) Refractive Index: 1.4630 Boiling Point: 98-100° Beilstein Reference: 1098377 UN No.: 1993 MDL No.: MFCD00000258

Formula: C₄H₇Br
Boiling Pt: 98…100 °C
Density: 1.330
Flash Pt: 1 °C (34 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000258
CAS Number: 5162-44-7
EINECS: 225-937-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR