(4-Hydroxyphenyl)acetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

A10971-03 A10971-06 A10971-14
CAAAA10971-03EA 52.46 CAD
CAAAA10971-03 CAAAA10971-06 CAAAA10971-14
(4-Hydroxyphenyl)acetonitrile 97%
(4-Hydroxyphenyl)acetonitrile

MDL: MFCD00002383 Beilstein Registry No.: 1934470


Purity: 97% CAS No.: 14191-95-8 Molecular Formula: C8H7NO Formula Weight: 133.15 Melting Point: 66-71° Boiling Point: 329-331° MDL No.: MFCD00002383

Formula: C₈H₇NO
Boiling Pt: 329…331 °C
Melting Pt: 66…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002383
CAS Number: 14191-95-8
EINECS: 238-046-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR