Acetohydrazide 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic hydrazide, Acethydrazide

A10969-14 A10969-22 A10969-36
CAAAA10969-14EA 98.28 CAD
CAAAA10969-14 CAAAA10969-22 CAAAA10969-36
Acetohydrazide 96%
Acetohydrazide

MDL: MFCD00007610 Beilstein Registry No.: 506165


Purity: 96% CAS No.: 1068-57-1 Molecular Formula: CH3CONHNH2 Formula Weight: 74.08 Flash Point: 110°(230°F) Melting Point: ca 60-68° Boiling Point: 128-129°/18mm Beilstein Reference: 506165 MDL No.: MFCD00007610

Formula: C₂H₆N₂O
Boiling Pt: 128…129 °C/18 mm
Melting Pt: ∼ 60…68 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007610
CAS Number: 1068-57-1
EINECS: 213-948-7
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR