(R)-(+)-α-Methylbenzylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-(+)-α-Phenylethylamine, (R)-(+)-1-Phenylethylamine, D(+)-α-Methylbenzylamine
MDL: MFCD00064405
Beilstein Registry No.: 2410916
Fieser: 1,838 3,199 5,441 6,457 11,411 12,319 13,185 14,257 17,257
Optical Rotation: +29°C (c=10 in ethanol)
Purity: 98%
CAS No.: 3886-69-9
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 71°(159°F)
Refractive Index: 1.5260
Odor: Amine-like
Melting Point: -10°
Boiling Point: 187°
Beilstein Reference: 2410916
Merck Reference: 14,6026
Optical Rotation: +29° (c=10 in ethanol)
UN No.: 2735
MDL No.: MFCD00064405
Formula:
C₈H₁₁N Boiling Pt: 187 °C Melting Pt: –10 °C Density: 0.948 Flash Pt: 71 °C (159 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064405 CAS Number: 3886-69-9 EINECS: 223-423-4 UN: 2922 ADR: 8,III Merck Index: 14,6026 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...