1,4-Naphthoquinone, (max. 5% H₂O) 97+% (dry weight)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,4-Naphthalenedione, 1,4-Dihydro-1,4-diketonaphthalene

A10958-0E A10958-22 A10958-36
CAAAA10958-0EEA 559.32 CAD
CAAAA10958-0E CAAAA10958-22 CAAAA10958-36
1,4-Naphthoquinone, (max. 5% H₂O) 97+% (dry weight)
1,4-Naphthoquinone

MDL: MFCD00001676 Beilstein Registry No.: 878524 Fieser: 1,714


Purity: 97+% CAS No.: 130-15-4 Molecular Formula: C10H6O2 Formula Weight: 158.15 Flash Point: 142°(288°F) Odor: Irritating Beilstein Reference: 878524 Merck Reference: 14,6395 UN No.: 2811 MDL No.: MFCD00001676

Formula: C₁₀H₆O₂
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 120…124 °C
Density: 1.420
Flash Pt: 142 °C (288 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001676
CAS Number: 130-15-4
EINECS: 204-977-6
UN: 2928
ADR: 6.1,II
Merck Index: 14,6395

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR