1,1,2,2-Tetrafluoro-1,2-diiodoethane 96% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with copper
A10954-06
A10954-03
CAAAA10954-06EA
320.74
CAD
CAAAA10954-06
CAAAA10954-03
1,1,2,2-Tetrafluoro-1,2-diiodoethane 96% stabilized
1,1,2,2-Tetrafluoro-1,2-diiodoethane
MDL: MFCD00039401
Beilstein Registry No.: 1740350
Purity: 96%
CAS No.: 354-65-4
Molecular Formula: IF2CCF2I
Formula Weight: 353.82
Flash Point: None
Refractive Index: 1.4950
Melting Point: -21°
Boiling Point: 112-113°
Beilstein Reference: 1740350
MDL No.: MFCD00039401
Formula:
C₂F₄I₂ Boiling Pt: 112…113 °C Melting Pt: –21 °C Density: 2.629 Flash Pt: None Storage Temperature: Ambient |
MDL Number:
MFCD00039401 CAS Number: 354-65-4 EINECS: 206-567-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...