(3-Methoxyphenyl)acetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10951-06
A10951-14
A10951-22
CAAAA10951-06EA
45.98
CAD
CAAAA10951-06
CAAAA10951-14
CAAAA10951-22
(3-Methoxyphenyl)acetic acid 97%
(3-Methoxyphenyl)acetic acid
MDL: MFCD00004334
Beilstein Registry No.: 2614004
Purity: 97%
CAS No.: 1798-09-0
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 67-72°
Beilstein Reference: 2614004
Formula:
(C₆H₄(OCH₃))CH₂COOH Boiling Pt: 305.95 °C (1013 hPa) Melting Pt: 67…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004334 CAS Number: 1798-09-0 EINECS: 217-282-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...