6-Nitroquinaldine 98%

Supplier: Thermo Scientific Chemicals
Warning

A10945-14 A10945-03 A10945-06
CAAAAA10945-14EA 1730.1 CAD
CAAAAA10945-14 CAAAA10945-03 CAAAA10945-06
6-Nitroquinaldine 98%
6-Nitroquinaldine

MDL: MFCD00051736 Beilstein Registry No.: 7915


Purity: 98% CAS No.: 613-30-9 Molecular Formula: C10H8N2O2 Formula Weight: 188.19 Melting Point: 162-166° Beilstein Reference: 7915

Formula: C₁₀H₈N₂O₂
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00051736
CAS Number: 613-30-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR