DL-Camphor 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00074738
Beilstein Registry No.: 1907611
Soluble in alcohols, ether, chloroform, benzene
Purity: 96%
CAS No.: 76-22-2
Molecular Formula: C10H16O
Formula Weight: 152.24
Flash Point: 64°(147°F)
Physical Form: Crystalline powder or lumps
Density: 0.992
Appearance: White
Odor: Camphor-like
Melting Point: 172-176°
Boiling Point: 204°
Solubility: Soluble in alcohols, ether, chloroform, benzene
Beilstein Reference: 1907611
Merck Reference: 14,1732
UN No.: 2717
MDL No.: MFCD00074738
Formula:
C₁₀H₁₆O Boiling Pt: 204 °C Melting Pt: 172…176 °C Density: 0.992 Flash Pt: 64 °C (147 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00074738 CAS Number: 76-22-2 EINECS: 200-945-0 UN: 2717 ADR: 4.1,III Merck Index: 14,1732 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...