1-Butyl isothiocyanate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10935-14
A10935-22
CAAAA10935-14EA
155.63
CAD
CAAAA10935-14
CAAAA10935-22
1-Butyl isothiocyanate 98+%
1-Butyl isothiocyanate
MDL: MFCD00004824
Beilstein Registry No.: 906839
Purity: 98+%
CAS No.: 592-82-5
Molecular Formula: C5H9NS
Formula Weight: 115.20
Flash Point: 66°(150°F)
Refractive Index: 1.5000
Odor: Pungent
Boiling Point: 63-65°/11mm
Beilstein Reference: 906839
UN No.: 2922
MDL No.: MFCD00004824
Formula:
C₅H₉NS Boiling Pt: 63…65 °C/11 mm Density: 0.954 Flash Pt: 66 °C (150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004824 CAS Number: 592-82-5 EINECS: 209-770-4 UN: 2927 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...