Azomethine H sodium salt hydrate
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10927-03
A10927-06
A10927-14
CAAAA10927-03EA
54.35
CAD
CAAAA10927-03
CAAAA10927-06
CAAAA10927-14
Azomethine H sodium salt hydrate
Azomethine H sodium salt hydrate
MDL: MFCD00149588
Beilstein Registry No.: 4100175
CAS No.: 206752-32-1
Molecular Formula: C17H12NNaO8S2.xH2O
Formula Weight: 445.41(anhy)
Melting Point:
Formula:
C₁₇H₁₂NNaO₈S₂·xH₂O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149588 CAS Number: 206752-32-1 EINECS: 250-975-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...