4-tert-Butylaniline 99%

Supplier: Thermo Scientific Chemicals
Danger

A10925-22 A10925-06 A10925-14
CAAAAA10925-22EA 835.82 CAD
CAAAAA10925-22 CAAAA10925-06 CAAAA10925-14
4-tert-Butylaniline 99%
4-tert-Butylaniline

MDL: MFCD00007899 Beilstein Registry No.: 2205786


Purity: 99% CAS No.: 769-92-6 Molecular Formula: C10H15N Formula Weight: 149.23 Flash Point: 101°(213°F) Refractive Index: 1.5380 Melting Point: 15-16° Boiling Point: 235° Beilstein Reference: 2205786 MDL No.: MFCD00007899

Formula: C₁₀H₁₅N
Boiling Pt: 235 °C
Melting Pt: 15…16 °C
Density: 0.941
Flash Pt: 101 °C (213 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007899
CAS Number: 769-92-6
EINECS: 212-215-9
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR