6-Fluoroanthranilic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10919-06
A10919-03
A10919-14
CAAAAA10919-06EA
199.08
CAD
CAAAAA10919-06
CAAAA10919-03
CAAAA10919-14
6-Fluoroanthranilic acid 98%
6-Fluoroanthranilic acid
MDL: MFCD00067781
Beilstein Registry No.: 3540926
Purity: 98%
CAS No.: 434-76-4
Molecular Formula: C7H6FNO2
Formula Weight: 155.13
Melting Point: 165-167°
Beilstein Reference: 3540926
Formula:
C₇H₆FNO₂ Boiling Pt: 305 °C (1013 hPa) Melting Pt: 165…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067781 CAS Number: 434-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...