2,3-Dimethylaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10918-0E
A10918-36
CAAAA10918-0EEA
499.28
CAD
CAAAA10918-0E
CAAAA10918-36
2,3-Dimethylaniline 98%
2,3-Dimethylaniline
MDL: MFCD00007732
Beilstein Registry No.: 742174
Purity: 98%
CAS No.: 87-59-2
Molecular Formula: C8H11N
Formula Weight: 121.18
Flash Point: 96°(204°F)
Refractive Index: 1.5680
Odor: Characteristic
Melting Point: 2-3°
Boiling Point: 221-222°
Beilstein Reference: 742174
Merck Reference: 14,10084
UN No.: 1711
MDL No.: MFCD00007732
Formula:
C₈H₁₁N Boiling Pt: 221…222 °C Melting Pt: 2…3 °C Density: 0.992 Flash Pt: 96 °C (204 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007732 CAS Number: 87-59-2 EINECS: 201-755-0 UN: 1711 ADR: 6.1,II Merck Index: 14,10084 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...